CAS RN 913738-04-2
CAS RN 913738-04-2
IUPAC Name:
9-[12-(9H-carbazol-9-yl)-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-4-yl]-9H-carbazole
Synonyms:
9-[12-(carbazol-9-yl)-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-4-yl]carbazole
Molecular Weight (g/mol):
514.65
Molecular Formula:
C36H22N2S
InChi Key:
SDHNJSIZTIODFW-UHFFFAOYSA-N
SMILES:
S1C2=C(C=C(C=C2)N2C3=C(C=CC=C3)C3=C2C=CC=C3)C2=C1C=CC(=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
2,8-Bis(9H-carbazol-9-yl)dibenzothiophene 98.0+%, TCI America™
CAS: 913738-04-2 Molecular Formula: C36H22N2S Molecular Weight (g/mol): 514.646 MDL Number: MFCD28291905 InChI Key: SDHNJSIZTIODFW-UHFFFAOYSA-N Synonym: DCzDBT, 2,8-Di(9-carbazolyl)dibenzothiophene PubChem CID: 59348313 IUPAC Name: 9-(8-carbazol-9-yldibenzothiophen-2-yl)carbazole SMILES: C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC5=C(C=C4)SC6=C5C=C(C=C6)N7C8=CC=CC=C8C9=CC=CC=C97