missing translation for 'onlineSavingsMsg'
Learn More

2,8-Bis(9H-carbazol-9-yl)dibenzothiophene 98.0+%, TCI America™

Catalog No. B4942200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4942200MG 200 mg
B49421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4942200MG Supplier TCI America Supplier No. B4942200MG
Only null left

Chemical Identifiers

CAS 913738-04-2
Molecular Formula C36H22N2S
Molecular Weight (g/mol) 514.646
MDL Number MFCD28291905
InChI Key SDHNJSIZTIODFW-UHFFFAOYSA-N
Synonym DCzDBT, 2,8-Di(9-carbazolyl)dibenzothiophene
PubChem CID 59348313
IUPAC Name 9-(8-carbazol-9-yldibenzothiophen-2-yl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC5=C(C=C4)SC6=C5C=C(C=C6)N7C8=CC=CC=C8C9=CC=CC=C97

Specifications

Melting Point 301°C
Color White-Yellow
Quantity 200 mg
Formula Weight 514.65
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,8-Bis(9H-carbazol-9-yl)dibenzothiophene