accessibility menu, dialog, popup

UserName

CAS RN 91296-87-6

IUPAC Name: hydrogen 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid chloride
Synonyms: sarafloxacin
Molecular Weight (g/mol): 421.83
Molecular Formula: C20H18ClF2N3O3
InChi Key: KNWODGJQLCISLC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC(=O)C1=CN(C2=CC=C(F)C=C2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)