missing translation for 'onlineSavingsMsg'
Learn More

Sarafloxacin Hydrochloride 97.0+%, TCI America™

Catalog No. S08401G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08401G Supplier TCI America Supplier No. S08401G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91296-87-6
Molecular Formula C20H18ClF2N3O3
Molecular Weight (g/mol) 421.829
MDL Number MFCD11869789
InChI Key KNWODGJQLCISLC-UHFFFAOYSA-N
Synonym sarafloxacin hydrochloride, sarafloxacin hcl, sarafloxacin hydrochloride usan, saraflox, sarafloxacin hydrochloride usan, saraflox, dsstox_cid_25939, dsstox_rid_81237
PubChem CID 56207
IUPAC Name 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;hydrochloride
SMILES C1CN(CCN1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl

Specifications

Quantity 1 g
Formula Weight 421.83
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sarafloxacin Hydrochloride
TSCA No
Recommended Storage Refrigerator