accessibility menu, dialog, popup

UserName

CAS RN 91296-86-5

IUPAC Name: hydrogen 6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid chloride
Synonyms: hydrogen difloxacin chloride
Molecular Weight (g/mol): 435.86
Molecular Formula: C21H20ClF2N3O3
InChi Key: JFMGBGLSDVIOHL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1CCN(CC1)C1=C(F)C=C2C(=C1)N(C=C(C(O)=O)C2=O)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results