missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Difloxacin hydrochloride, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 91296-86-5 |
| Physical Form | Neat |
| Linear Formula | C21H19F2N3O3 · HCl |
| MDL Number | MFCD03840489 |
| Synonym | 6-Fluoro-1-(4-fluorophenyl)-1,4-dihydro-7-(4-methylpiperazino)-4-oxo-3-quinolinecarboxylic acid hydrochloride |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 435.85 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH