accessibility menu, dialog, popup

UserName

CH3NNHOO

CAS RN 91138-00-0

IUPAC Name: 5-methyl-1-phenyl-1H-pyrazole-4-carboxylic acid
Synonyms: 5-methyl-1-phenylpyrazole-4-carboxylic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: USSMIQWDLWJQDQ-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1C1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)