missing translation for 'onlineSavingsMsg'
Learn More

TAPS sodium salt, 98%

Catalog No. AAJ6118922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6118922 100 g
AAJ6118914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6118922 Supplier Thermo Scientific Chemicals Supplier No. J6118922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Zwitterionic buffer | CAS: 91000-53-2 | C7H16NNaO6S | 265.256 g/mol

TAPS sodium salt is used in biochemistry and molecular biology. It has a useful pH range of 7.7 - 9.1. It has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. It has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TAPS sodium salt is used in biochemistry and molecular biology. It has a useful pH range of 7.7 - 9.1. It has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. It has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

Solubility
Soluble in water (333.33 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 91000-53-2
Molecular Formula C7H16NNaO6S
Molecular Weight (g/mol) 265.256
MDL Number MFCD00069903
InChI Key FEGYIWVHCSRXCG-UHFFFAOYSA-M
PubChem CID 23667519
IUPAC Name sodium;3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonate
SMILES C(CNC(CO)(CO)CO)CS(=O)(=O)[O-].[Na+]

Specifications

Color White
Physical Form Powder
Quantity 100 g
Linear Formula (HOCH2)3CNH(CH2)3SO3Na
Solubility Information Soluble in water (333.33mg/ml).
Formula Weight 265.26
Percent Purity 98%
Chemical Name or Material TAPS sodium salt
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only