accessibility menu, dialog, popup

UserName

CAS RN 91-96-3

IUPAC Name: N-[3,3'-dimethyl-4'-(3-oxobutanamido)-[1,1'-biphenyl]-4-yl]-3-oxobutanamide
Synonyms: C.I. azoic coupling component 5
Molecular Weight (g/mol): 380.44
Molecular Formula: C22H24N2O4
InChi Key: CRRLDLPBQWRVGN-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC=C(C=C1C)C1=CC=C(NC(=O)CC(C)=O)C(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results