missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(acetoacetyl)-o-toluidine 95.0+%, TCI America™

Catalog No. B078825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B078825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B078825G Supplier TCI America Supplier No. B078825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91-96-3
Molecular Formula C22H24N2O4
Molecular Weight (g/mol) 380.44
MDL Number MFCD00026248
InChI Key CRRLDLPBQWRVGN-UHFFFAOYSA-N
Synonym N,N′C-Bis(acetoacetyl)-3,3′C-dimethylbenzidine, Naphthol AS-G, Azoic Coupling Component 5
PubChem CID 66686
IUPAC Name N-[3,3'-dimethyl-4'-(3-oxobutanamido)-[1,1'-biphenyl]-4-yl]-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=C(C=C1C)C1=CC=C(NC(=O)CC(C)=O)C(C)=C1

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 25 g
Formula Weight 380.44
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(acetoacetyl)-o-toluidine
EINECSNumber (5)-2275
TSCA Yes