accessibility menu, dialog, popup

UserName

CAS RN 91-13-4

IUPAC Name: 1,2-bis(bromomethyl)benzene
Synonyms: benzene, 1,2-bis(bromomethyl)-
Molecular Weight (g/mol): 263.96
Molecular Formula: C8H8Br2
InChi Key: KGKAYWMGPDWLQZ-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC=C1CBr
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)