missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dibromo-o-xylene, 96%

Catalog No. AC112931000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112931000 100 g Plastic bottle
AC112930250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112931000 Supplier Thermo Scientific Chemicals Supplier No. 112931000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91-13-4 | C8H8Br2 | 263.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91-13-4
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.95
MDL Number MFCD00000175
InChI Key KGKAYWMGPDWLQZ-UHFFFAOYSA-N
Synonym 1,2-bis bromomethyl benzene, o-xylylene dibromide, alpha,alpha'-dibromo-o-xylene, benzene, 1,2-bis bromomethyl, o-bis bromomethyl benzene, o-xylylene bromide, 1,2-dibenzyl bromide, alpha,alpha'-dibromo-o-xylol, a,a'-dibromo-o-xylene, ccris 1776
PubChem CID 66665
IUPAC Name 1,2-bis(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)CBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 91.0°C to 96.0°C
Color White to Cream
Density 1.9600g/mL
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic bottle
Linear Formula C6H4(CH2Br)2
Quantity 100 g
Beilstein 05, 366
Specific Gravity 1.96
Solubility Information Solubility in water: soluble,hydrolyses. Other solubilities: soluble in ethanol
Formula Weight 263.95
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material α, α'-Dibromo-o-xylene, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-042-7
TSCA TSCA

RUO – Research Use Only