CAS RN 9067-32-7
Sodium hyaluronate, 95%
CAS: 9067-32-7 Molecular Formula: (C14H20NO11Na)n Molecular Weight (g/mol): 417.30 MDL Number: MFCD00875848 InChI Key: YWIVKILSMZOHHF-QJZPQSOGSA-N Synonym: hyaluronate tetrasaccharide,d0e9sz,6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid IUPAC Name: Sodium hyaluronate SMILES: CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*
MilliporeSigma™ Hyaluronic Acid, Sodium Salt, Streptococcus pyrogenes, Calbiochem™,
Molecular Formula: (C14H20NO11Na)n Molecular Weight (g/mol): 417.3 MDL Number: MFCD00875848 InChI Key: YWIVKILSMZOHHF-QJZPQSOGSA-N Synonym: hyaluronate tetrasaccharide,d0e9sz,6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid IUPAC Name: Sodium hyaluronate SMILES: CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*
Hyaluronic acid sodium salt, Streptococcus equi, 91%
CAS: 9067-32-7 Molecular Formula: (C14H20NO11Na)n Molecular Weight (g/mol): 417.30 MDL Number: MFCD00875848 InChI Key: YWIVKILSMZOHHF-QJZPQSOGSA-N IUPAC Name: Sodium hyaluronate SMILES: CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*