missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Hyaluronic Acid, Sodium Salt, Streptococcus pyrogenes, Calbiochem™,

Catalog No. 385908500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
385908500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 385908500MG Supplier MilliporeSigma™ Supplier No. 385908500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

Molecular Formula (C14H20NO11Na)n
Molecular Weight (g/mol) 417.3
MDL Number MFCD00875848
InChI Key YWIVKILSMZOHHF-QJZPQSOGSA-N
Synonym hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
IUPAC Name Sodium hyaluronate
SMILES CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*

Specifications

Packaging Glass Bottle
Physical Form Lyophilized
Quantity 500 mg