missing translation for 'onlineSavingsMsg'
Learn More

Sodium hyaluronate, 95%

Catalog No. AC251770250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC251770250 25 g Glass bottle
AC251770010 1 g Glass bottle
AC251770050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC251770250 Supplier Thermo Scientific Chemicals Supplier No. 251770250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9067-32-7
Molecular Formula (C14H20NO11Na)n
Molecular Weight (g/mol) 417.30
MDL Number MFCD00875848
InChI Key YWIVKILSMZOHHF-QJZPQSOGSA-N
Synonym hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
IUPAC Name Sodium hyaluronate
SMILES CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*

Specifications

Color White to Cream
pH 6.0 to 8.0 (0.5% aq. soln.)
Loss on Drying 10% max. (500mg, 105°C, 6 hrs)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (On dry substance)
Packaging Glass bottle
Linear Formula (C14H20NO11Na)n
Quantity 25 g
Merck Index 15,4796
Solubility Information Solubility in water: soluble.
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Sodium hyaluronate
Show More Show Less

RUO – Research Use Only