accessibility menu, dialog, popup

UserName

CAS RN 904326-92-7

BNFCH3HOOH

CAS RN 904326-92-7

IUPAC Name: (6-fluoro-5-methylpyridin-3-yl)boronic acid
Synonyms: 6-fluoro-5-methylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 154.94
Molecular Formula: C6H7BFNO2
InChi Key: XCWIMBMPJGQNHG-UHFFFAOYSA-N
SMILES: CC1=C(F)N=CC(=C1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results