missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methylpyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F09841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09841G Supplier TCI America Supplier No. F09841G
Only null left

Chemical Identifiers

CAS 904326-92-7
Molecular Formula C6H7BFNO2
Molecular Weight (g/mol) 154.935
MDL Number MFCD03411562
InChI Key XCWIMBMPJGQNHG-UHFFFAOYSA-N
Synonym 2-fluoro-3-methylpyridine-5-boronic acid, 6-fluoro-5-methylpyridin-3-yl boronic acid, 2-fluoro-3-methylpyridine-5-yl boronic acid, 2-fluoro-3-methyl-5-pyridineboronic acid, 6-fluoro-5-methylpyridine-3-boronic acid, 6-fluoro-5-methylpyridin-3-yl-3-boronic acid, 2-fluoro-3-picoline-5-boronic acid, 6-fluoro-5-methylpyridine-3-boronicacid, 6-fluoro-5-methyl-3-pyridineboronic acid, 6-fluoro-5-methyl-3-pyridyl boronic acid
PubChem CID 17750247
IUPAC Name (6-fluoro-5-methylpyridin-3-yl)boronic acid
SMILES B(C1=CC(=C(N=C1)F)C)(O)O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 154.94
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-3-methylpyridine-5-boronic Acid (contains varying amounts of Anhydride)