accessibility menu, dialog, popup

UserName

CAS RN 9036-66-2

IUPAC Name: 4-[(6-{[(3,5-dihydroxy-4-methoxyoxan-2-yl)oxy]methyl}-3,5-dihydroxy-4-methoxyoxan-2-yl)oxy]-2-(hydroxymethyl)-6-methyloxane-3,5-diol
Synonyms: 4-[(6-{[(3,5-dihydroxy-4-methoxyoxan-2-yl)oxy]methyl}-3,5-dihydroxy-4-methoxyoxan-2-yl)oxy]-2-(hydroxymethyl)-6-methyloxane-3,5-diol galactoarabinan
Molecular Weight (g/mol): 500.49
Molecular Formula: C20H36O14
InChi Key: SATHPVQTSSUFFW-UHFFFAOYNA-N
SMILES: COC1C(O)COC(OCC2OC(OC3C(O)C(C)OC(CO)C3O)C(O)C(OC)C2O)C1O
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results