missing translation for 'onlineSavingsMsg'
Learn More

(+)-Arabinogalactan from Larch Wood, TCI America™

Catalog No. A132825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A132825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A132825G Supplier TCI America Supplier No. A132825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9036-66-2
Molecular Formula C20H36O14
Molecular Weight (g/mol) 500.494
MDL Number MFCD00062638
InChI Key SATHPVQTSSUFFW-UHFFFAOYSA-N
Synonym Polyarabinogalactan
PubChem CID 24847856
IUPAC Name 4-[6-[(3,5-dihydroxy-4-methoxyoxan-2-yl)oxymethyl]-3,5-dihydroxy-4-methoxyoxan-2-yl]oxy-2-(hydroxymethyl)-6-methyloxane-3,5-diol
SMILES CC1C(C(C(C(O1)CO)O)OC2C(C(C(C(O2)COC3C(C(C(CO3)O)OC)O)O)OC)O)O

Specifications

Physical Form Crystal-Powder at 20°C
Quantity 25 g
Chemical Name or Material (+)-Arabinogalactan from Larch Wood