accessibility menu, dialog, popup

UserName

CAS RN 9007-49-2

A(a)

CAS RN 9007-49-2

IUPAC Name: Deoxyribonucleic acid
Synonyms:
Molecular Weight (g/mol): 0
Molecular Formula:
InChi Key:
SMILES: *
Form 
  • (2)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results