accessibility menu, dialog, popup

UserName

CAS RN 9007-28-7

CH3ONH(R)(R)OHO(R)OSHOOO(R)HO(R)O(R)O(S)(S)HO(S)HO(R)OHHOO

CAS RN 9007-28-7

IUPAC Name: (2S,3S,4S,5R,6R)-6-{[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-(sulfooxy)oxan-4-yl]oxy}-3,4,5-trihydroxyoxane-2-carboxylic acid
Synonyms: (2S,3S,4S,5R,6R)-6-{[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-(sulfooxy)oxan-4-yl]oxy}-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular Weight (g/mol): 463.36
Molecular Formula: C13H21NO15S
InChi Key: KXKPYJOVDUMHGS-OSRGNVMNSA-N
SMILES: CC(=O)N[C@H]1[C@H](O)O[C@H](OS(O)(=O)=O)[C@H](O)[C@@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results