missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Chondroitin Sulfate, Bovine Trachea

Catalog No. 2306995GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2306995GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2306995GM Supplier MilliporeSigma™ Supplier No. 2306995GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fundamental component of connective tissue

  • Fundamental component of connective tissue
  • Acts as a flexible connecting matrix between tough protein filaments in cartilage to form a polymeric system
  • Mixture of chondroitin sulfates A, C, D, and E

Specifications

CAS 9007-28-7
Content And Storage Store at +2°C to +8°C
Formula Weight 50000g/mol
Purity ≥90% by HPLC
Quantity 5 g
Synonym Chondroitin Polysulfate, Chonsurid
Form Solid