accessibility menu, dialog, popup

UserName

CAS RN 90069-81-1

IUPAC Name: thiophene-3,4-diamine dihydrochloride
Synonyms: thiophene-3,4-diamine dihydrochloride
Molecular Weight (g/mol): 187.08
Molecular Formula: C4H8Cl2N2S
InChi Key: RAMOMCXNLLLICQ-UHFFFAOYSA-N
SMILES: Cl.Cl.NC1=CSC=C1N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)