missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminothiophene dihydrochloride, 96%

Catalog No. AC325890010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325890010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC325890010 Supplier Thermo Scientific Chemicals Supplier No. 325890010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90069-81-1 | C4H6N2S·2HCl | 187.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90069-81-1
Molecular Formula C4H6N2S·2HCl
Molecular Weight (g/mol) 187.09
MDL Number MFCD00671511
InChI Key RAMOMCXNLLLICQ-UHFFFAOYSA-N
Synonym 3,4-diaminothiophene dihydrochloride, thiophene-3,4-diamine dihydrochloride, 3,4-diaminothiophenedihydrochloride, pubchem21822, acmc-209r4q, 3,4-diaminothiophene 2hcl, 3?4-diaminothiophene dihydrochloride, 3,4-thiophenediamine, dihydrochloride, thiophene-3,4-diamine-hydrogen chloride 1/2
PubChem CID 12707342
IUPAC Name thiophene-3,4-diamine;dihydrochloride
SMILES C1=C(C(=CS1)N)N.Cl.Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 240.0°C to 245.0°C
Color Brown or Gray
Loss on Drying 3% max.
Infrared Spectrum Authentic
Assay Percent Range 95% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Quantity 1 g
Formula Weight 187.09
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Diaminothiophene dihydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
TSCA TSCA

RUO – Research Use Only