CAS RN 9000-70-8
CAS RN 9000-70-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Gelatin Type A Type A; 225 Bloom Food Grade MP Biomedicals
CAS: 9000-70-8 MDL Number: MFCD00081638
2
Gelatin, 250 Bloom, Powder, NF, Spectrum™ Chemical
CAS: 9000-70-8 MDL Number: MFCD00081638