CAS RN 90-72-2
IUPAC Name:
2,4,6-tris[(dimethylamino)methyl]phenol
Synonyms:
2,4,6-tris[(dimethylamino)methyl]phenol
Molecular Weight (g/mol):
265.4
Molecular Formula:
C15H27N3O
InChi Key:
AHDSRXYHVZECER-UHFFFAOYSA-N
SMILES:
CN(C)CC1=CC(CN(C)C)=C(O)C(CN(C)C)=C1
1
–
1
of
1
results
1
2,4,6-Tris(dimethylaminomethyl)phenol 80.0+%, TCI America™
CAS: 90-72-2 Molecular Formula: C15H27N3O Molecular Weight (g/mol): 265.401 MDL Number: MFCD00008330 InChI Key: AHDSRXYHVZECER-UHFFFAOYSA-N Synonym: 2,4,6-tris dimethylaminomethyl phenol,sumicure d,2,4,6-tris dimethylamino methyl phenol,dmf 3,actiron nx 3,ancamine k 54,capcure eh 30,epilink 230,araldite dy 061 PubChem CID: 7026 IUPAC Name: 2,4,6-tris[(dimethylamino)methyl]phenol SMILES: CN(C)CC1=CC(=C(C(=C1)CN(C)C)O)CN(C)C