missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(dimethylaminomethyl)phenol 80.0+%, TCI America™

Catalog No. T0526500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0526500G 500 g
T052625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0526500G Supplier TCI America Supplier No. T0526500G

Chemical Identifiers

CAS 90-72-2
Molecular Formula C15H27N3O
Molecular Weight (g/mol) 265.401
MDL Number MFCD00008330
InChI Key AHDSRXYHVZECER-UHFFFAOYSA-N
Synonym 2,4,6-tris dimethylaminomethyl phenol, sumicure d, 2,4,6-tris dimethylamino methyl phenol, dmf 3, actiron nx 3, ancamine k 54, capcure eh 30, epilink 230, araldite dy 061
PubChem CID 7026
IUPAC Name 2,4,6-tris[(dimethylamino)methyl]phenol
SMILES CN(C)CC1=CC(=C(C(=C1)CN(C)C)O)CN(C)C

Specifications

Color Yellow
Boiling Point 135°C
Quantity 500 g
UN Number 1760
Formula Weight 265.40
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6-Tris(dimethylaminomethyl)phenol
EINECSNumber (3)-0762&(3)-0776
RTECSNumber SN3500000
TSCA Yes