man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 898544-50-8

BNNNFFFK

CAS RN 898544-50-8

IUPAC Name: potassium [4-(azidomethyl)phenyl]trifluoroboranuide
Synonyms: potassium [4-(azidomethyl)phenyl]trifluoroboranuide
Molecular Weight (g/mol): 239.05
Molecular Formula: C7H6BF3KN3
InChi Key: XIWGKBHBLASHCL-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(CN=[N+]=[N-])C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results