accessibility menu, dialog, popup

UserName

CAS RN 898289-18-4

CH3ONCO

CAS RN 898289-18-4

IUPAC Name: 3-isocyanato-5-methyl-2-phenylfuran
Synonyms: 3-isocyanato-5-methyl-2-phenylfuran
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: GDLFJANXKGIZKA-UHFFFAOYSA-N
SMILES: CC1=CC(N=C=O)=C(O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)