missing translation for 'onlineSavingsMsg'
Learn More

3-Isocyanato-5-methyl-2-phenylfuran, 97%, Thermo Scientific™

Chemical Identifiers

CAS 898289-18-4
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD09025851
InChI Key GDLFJANXKGIZKA-UHFFFAOYSA-N
Synonym 5-methyl-2-phenylfuran-3-isocyanate, furan,3-isocyanato-5-methyl-2-phenyl
PubChem CID 18525836
IUPAC Name 3-isocyanato-5-methyl-2-phenylfuran
SMILES CC1=CC(=C(O1)C2=CC=CC=C2)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC49706CB
SDS View Documents
Thermo Scientific Chemicals
CC49706CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC49706DA
SDS View Documents
Thermo Scientific Chemicals
CC49706DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

898289-18-4
199.209
GDLFJANXKGIZKA-UHFFFAOYSA-N
18525836
CC1=CC(=C(O1)C2=CC=CC=C2)N=C=O
C12H9NO2
MFCD09025851
5-methyl-2-phenylfuran-3-isocyanate, furan,3-isocyanato-5-methyl-2-phenyl
3-isocyanato-5-methyl-2-phenylfuran

Specifications

898289-18-4
100.0
Amber Glass Bottle
MFCD09025851
5-methyl-2-phenylfuran-3-isocyanate, furan,3-isocyanato-5-methyl-2-phenyl
CC1=CC(=C(O1)C2=CC=CC=C2)N=C=O
199.209
199.21
3-Isocyanato-5-methyl-2-phenylfuran
96.5
97%
C12H9NO2
250mg
GDLFJANXKGIZKA-UHFFFAOYSA-N
3-isocyanato-5-methyl-2-phenylfuran
18525836
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents