accessibility menu, dialog, popup

UserName

CAS RN 89808-01-5

CH3OH2NHCl

CAS RN 89808-01-5

IUPAC Name: 2-ethoxyaniline hydrochloride
Synonyms: 2-ethoxyaniline hydrochloride
Molecular Weight (g/mol): 173.64
Molecular Formula: C8H12ClNO
InChi Key: HODQIPYRQHAZFV-UHFFFAOYSA-N
SMILES: Cl.CCOC1=CC=CC=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results