missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Phenetidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P009225G
Specifications
| o-Phenetidine Hydrochloride | |
| Purple-White | |
| MFCD00060217 | |
| 2-Ethoxyaniline Hydrochloride | |
| CCOC1=CC=CC=C1N.Cl | |
| 173.64 | |
| 173.64 | |
| Crystalline Powder |
| 89808-01-5 | |
| C8H12ClNO | |
| 25 g | |
| HODQIPYRQHAZFV-UHFFFAOYSA-N | |
| 2-ethoxyaniline;hydrochloride | |
| 12526745 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 89808-01-5 | |
| 173.64 | |
| HODQIPYRQHAZFV-UHFFFAOYSA-N | |
| 12526745 | |
| CCOC1=CC=CC=C1N.Cl |
| C8H12ClNO | |
| MFCD00060217 | |
| 2-Ethoxyaniline Hydrochloride | |
| 2-ethoxyaniline;hydrochloride |
Safety and Handling
EINECSNumber : (3)-0682
TSCA : No