CAS RN 89797-68-2
IUPAC Name:
5-(ethylsulfanyl)-2H-1,2,3,4-tetrazole
Synonyms:
5-(ethylsulfanyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol):
130.17
Molecular Formula:
C3H6N4S
InChi Key:
GONFBOIJNUKKST-UHFFFAOYSA-N
SMILES:
CCSC1=NNN=N1
1
–
5
of
5
results
1
Activator Reagent, Solstice
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.25M ETT in Acetonitrile
2
Activator Reagent,Solstice
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.25M ETT in Acetonitrile
3
Activator Reagent, Solstice
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.25M ETT in Acetonitrile
4
Activator Reagent, Solstice
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.5M ETT in Acetonitrile
5
5-(Ethylthio)-1H-tetrazole 98.0+%, TCI America™
CAS: 89797-68-2 Molecular Formula: C3H6N4S Molecular Weight (g/mol): 130.169 MDL Number: MFCD00068733 InChI Key: GONFBOIJNUKKST-UHFFFAOYSA-N PubChem CID: 2758928 IUPAC Name: 5-ethylsulfanyl-2H-tetrazole SMILES: CCSC1=NNN=N1