missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activator Reagent, Solstice
Specifications
Specifications
| CAS | 89797-68-2, 75-05-8 |
| CAS Max % | 4.00%, 96.00% |
| Quantity | 4 x 2.5 L |
| Flash Point | 7°C |
| Packaging | GL45 Glass Bottle |
| Physical Form | Liquid |
| Purity Grade Notes | Activator reagents |
| Boiling Point | 82°C.The physical data is that of the main component. |