accessibility menu, dialog, popup

UserName

CAS RN 89642-49-9

IUPAC Name: 4-bromo-3-nitrobenzonitrile
Synonyms: 4-bromo-3-nitrobenzonitrile
Molecular Weight (g/mol): 227.02
Molecular Formula: C7H3BrN2O2
InChi Key: FXRMUJPWDOLCLX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1Br)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)