missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitrobenzonitrile 98.0+%, TCI America™

Catalog No. B47085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47085G 5 g
B47081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47085G Supplier TCI America Supplier No. B47085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89642-49-9
Molecular Formula C7H3BrN2O2
Molecular Weight (g/mol) 227.017
MDL Number MFCD00982006
InChI Key FXRMUJPWDOLCLX-UHFFFAOYSA-N
PubChem CID 3926861
IUPAC Name 4-bromo-3-nitrobenzonitrile
SMILES C1=CC(=C(C=C1C#N)[N+](=O)[O-])Br

Specifications

Melting Point 121°C
Quantity 5 g
Formula Weight 227.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-nitrobenzonitrile
TSCA No
Recommended Storage Refrigerator