accessibility menu, dialog, popup

UserName

HClOHONO

CAS RN 89531-58-8

IUPAC Name: hydrogen 2-(morpholin-4-yl)acetic acid chloride
Synonyms: hydrogen morpholin-4-ylacetic acid chloride
Molecular Weight (g/mol): 181.62
Molecular Formula: C6H12ClNO3
InChi Key: ICGYBJNONRDZOP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC(=O)CN1CCOCC1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)