missing translation for 'onlineSavingsMsg'
Learn More

Morpholinoacetic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. M26085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26085G 5 g
M26081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26085G Supplier TCI America Supplier No. M26085G

Chemical Identifiers

CAS 89531-58-8
Molecular Formula C6H12ClNO3
Molecular Weight (g/mol) 181.62
MDL Number MFCD03540888
InChI Key ICGYBJNONRDZOP-UHFFFAOYSA-N
Synonym 2-morpholinoacetic acid hydrochloride, 2-morpholin-4-yl acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholinylacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1
PubChem CID 17750946
IUPAC Name hydrogen 2-(morpholin-4-yl)acetic acid chloride
SMILES [H+].[Cl-].OC(=O)CN1CCOCC1

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 5 g
Formula Weight 181.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Morpholinoacetic Acid Hydrochloride