accessibility menu, dialog, popup

UserName

CAS RN 890-38-0

IUPAC Name: 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: 2'-deoxyinosine
Molecular Weight (g/mol): 252.23
Molecular Formula: C10H12N4O4
InChi Key: VGONTNSXDCQUGY-YUZWJPFSNA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)