accessibility menu, dialog, popup

UserName

CAS RN 89-73-6

IUPAC Name: N,2-dihydroxybenzamide
Synonyms: salicylhydroxamic acid salicyloylhydroxamic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: HBROZNQEVUILML-UHFFFAOYSA-N
SMILES: ONC(=O)C1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)