missing translation for 'onlineSavingsMsg'
Learn More

Salicylhydroxamic Acid 98.0+%, TCI America™

Catalog No. S00085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00085G 5 g
S000825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S00085G Supplier TCI America Supplier No. S00085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89-73-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00002110
InChI Key HBROZNQEVUILML-UHFFFAOYSA-N
Synonym salicylhydroxamic acid, 2-hydroxybenzohydroxamic acid, salicylohydroxamic acid, sham, salicylohydroximic acid, benzamide, n,2-dihydroxy, 2-hydroxybenzhydroxamic acid, o-hydroxybenzohydroxamic acid, sha, 2,n-dihydroxy-benzamide
PubChem CID 66644
ChEBI CHEBI:45615
IUPAC Name N,2-dihydroxybenzamide
SMILES C1=CC=C(C(=C1)C(=O)NO)O

Specifications

Melting Point 173°C
Quantity 5 g
Formula Weight 153.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Salicylhydroxamic Acid
RTECSNumber VO6870000
TSCA No