accessibility menu, dialog, popup

UserName

CAS RN 889940-13-0

IUPAC Name: 3,3,3-trifluoro-2,2-dimethylpropanoic acid
Synonyms: 3,3,3-trifluoro-2,2-dimethylpropanoic acid
Molecular Weight (g/mol): 156.1
Molecular Formula: C5H7F3O2
InChi Key: VMFSJVUPIXOCFO-UHFFFAOYSA-N
SMILES: CC(C)(C(O)=O)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)