missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-2,2-dimethylpropionic Acid 98.0+%, TCI America™

Catalog No. T34501G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T34501G Supplier TCI America Supplier No. T34501G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 889940-13-0
Molecular Formula C5H7F3O2
Molecular Weight (g/mol) 156.104
MDL Number MFCD08445819
InChI Key VMFSJVUPIXOCFO-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-2,2-dimethylpropionic acid, 2-trifluoromethyl-2-methylpropanoic acid, 3,3,3-trifluoro-2,2-dimethyl-propionic acid, 2-trifluoromethyl-isobutyric acid, 3,3,3-trifluoro-2,2-dimethyl-propanoic acid
PubChem CID 19867367
IUPAC Name 3,3,3-trifluoro-2,2-dimethylpropanoic acid
SMILES CC(C)(C(=O)O)C(F)(F)F

Specifications

Melting Point 73°C
Color White
Boiling Point 77°C
Quantity 1 g
UN Number 3261
Formula Weight 156.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,3-Trifluoro-2,2-dimethylpropionic Acid