CAS RN 88889-14-9
IUPAC Name:
sodium (2S,4S)-4-cyclohexyl-1-(2-{[2-methyl-1-(propanoyloxy)propoxy](4-phenylbutyl)phosphoryl}acetyl)pyrrolidine-2-carboxylate
Synonyms:
fosinopril sodium
Molecular Weight (g/mol):
585.65
Molecular Formula:
C30H45NNaO7P
InChi Key:
TVTJZMHAIQQZTL-HREVRLCXSA-M
SMILES:
[Na+].CCC(=O)OC(OP(=O)(CCCCC1=CC=CC=C1)CC(=O)N1C[C@@H](C[C@H]1C([O-])=O)C1CCCCC1)C(C)C
1
–
1
of
1
results
1
Thermo Scientific™ Fosinopril (sodium), 98%
CAS: 88889-14-9 Molecular Formula: C{3}{0}H{4}{5}NO{7}PNa