missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fosinopril (sodium), 98%

Catalog No. AAH3712003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3712003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3712003 Supplier Thermo Scientific™ Supplier No. H37120MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88889-14-9 | C{3}{0}H{4}{5}NO{7}PNa, C30H45NNaO7P | 585.65 g/mol

Chemical Identifiers

CAS 88889-14-9
Molecular Formula C{3}{0}H{4}{5}NO{7}PNa

Specifications

Quantity 1 g
Melting Point 190-192°
Formula Weight 585.65
Physical Form Solid
Purity 98%
EINECSNumber 620-449-1