accessibility menu, dialog, popup

UserName

CAS RN 888-54-0

ONOHNNCH2NOO(E)

CAS RN 888-54-0

IUPAC Name: (2E)-1-(2,4-dinitrophenyl)-2-(prop-2-en-1-ylidene)hydrazine
Synonyms: (2E)-1-(2,4-dinitrophenyl)-2-(prop-2-en-1-ylidene)hydrazine
Molecular Weight (g/mol): 236.19
Molecular Formula: C9H8N4O4
InChi Key: JJPZHGIYUVFTGG-BJMVGYQFSA-N
SMILES: [O-][N+](=O)C1=CC=C(N\N=C\C=C)C(=C1)[N+]([O-])=O
Concentration 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)