missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Acrolein-DNPH, 1,000 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111020376
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020376 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020376 Supplier Merck Emd Millipore Supplier No. CRM47342
Only null left

Specifications

Packaging Ampule
CAS 888-54-0
Product Line TraceCERT
Flash Point 2.0°C - closed cup
Concentration 1000 μg/mL in acetonitrile (as the aldehyde)
Linear Formula C9H8N4O4
Synonym Acrolein-2,4-dinitrophenylhydrazone solution; Acrolein-2,4-DNPH solution
Type Organic Pollutant Standard
Quantity 1 mL
Formula Weight 236.18
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC