accessibility menu, dialog, popup

UserName

CH3OOOSOOFFF

CAS RN 88767-98-0

IUPAC Name: ethyl 4-phenyl-2-(trifluoromethanesulfonyloxy)butanoate
Synonyms: ethyl 4-phenyl-2-(trifluoromethanesulfonyloxy)butanoate
Molecular Weight (g/mol): 340.31
Molecular Formula: C13H15F3O5S
InChi Key: UDJNHUAEPSYCRU-UHFFFAOYNA-N
SMILES: CCOC(=O)C(CCC1=CC=CC=C1)OS(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results