missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-4-Phenyl-2-(trifluoromethanesulfonyloxy)butyrate 97.0+%, TCI America™

Catalog No. E08791G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08791G Supplier TCI America Supplier No. E08791G
Only null left

Chemical Identifiers

CAS 88767-98-0
Molecular Formula C13H15F3O5S
Molecular Weight (g/mol) 340.313
MDL Number MFCD09751096
InChI Key UDJNHUAEPSYCRU-LLVKDONJSA-N
Synonym (R)-4-Phenyl-2-(trifluoromethanesulfonyloxy)butyric Acid Ethyl Ester
PubChem CID 10860745
IUPAC Name ethyl (2R)-4-phenyl-2-(trifluoromethylsulfonyloxy)butanoate
SMILES CCOC(=O)C(CCC1=CC=CC=C1)OS(=O)(=O)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 340.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (R)-4-Phenyl-2-(trifluoromethanesulfonyloxy)butyrate