accessibility menu, dialog, popup

UserName

H3COOOOOOCH3

CAS RN 88754-66-9

IUPAC Name: [({2-[(methoxycarbonyl)oxy]ethoxy}carbonyl)oxy]methane
Synonyms: [({2-[(methoxycarbonyl)oxy]ethoxy}carbonyl)oxy]methane
Molecular Weight (g/mol): 178.14
Molecular Formula: C6H10O6
InChi Key: DOMLQXFMDFZAAL-UHFFFAOYSA-N
SMILES: COC(=O)OCCOC(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results